Assay Details
| Gene Symbol: Nedd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111324.2 | NP_001104794.1 | 5 - 6 | 1012 | 136 | |
| NM_017464.5 | NP_059492.3 | 4 - 5 | 793 | 136 | ||
| GenBank mRNA | AF009366.1 | - | 5 - 6 | 948 | 136 | |
| AK030985.1 | - | 5 - 6 | 946 | 136 | ||
| AK033729.1 | - | 4 - 5 | 794 | 136 | ||
| AK046357.1 | - | 4 - 5 | 756 | 136 | ||
| AK054179.1 | - | 4 - 5 | 789 | 136 | ||
| AK083374.1 | - | 5 - 6 | 940 | 136 | ||
| BC053713.1 | - | 5 - 6 | 945 | 30077589 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nedd9
Genomic Map