Assay Details
| Gene Symbol: Dusp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024438.4 | NP_077758.1 | 2 - 3 | 441 | 136 | |
| GenBank mRNA | AB038769.1 | - | 2 - 3 | 396 | 136 | |
| AB051896.1 | - | 2 - 3 | 415 | 136 | ||
| AF237618.1 | - | 2 - 3 | 613 | 136 | ||
| AK010127.1 | - | 2 - 3 | 438 | 136 | ||
| AK018369.1 | - | 2 - 3 | 98 | 136 | ||
| AK077871.1 | - | 2 - 3 | 442 | 136 | ||
| BC021591.1 | - | 2 - 3 | 396 | 5149051 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dusp19
Genomic Map