Assay Details
| Gene Symbol: Mvb12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175184.4 | NP_780393.2 | 4 - 5 | 956 | 65 | |
| GenBank mRNA | AK012178.1 | - | 4 - 5 | 449 | 65 | |
| AK131113.1 | - | 4 - 5 | 474 | 65 | ||
| AK135990.1 | - | 4 - 5 | 390 | 65 | ||
| AK154267.1 | - | 4 - 5 | 450 | 65 | ||
| BC049129.1 | - | 4 - 5 | 469 | 6825067 | 65 | |
| BC059907.1 | - | 4 - 5 | 956 | 6824889 | 65 | |
More Information
Set Membership: |
|

Mvb12b
Genomic Map