Assay Details
| Gene Symbol: Jarid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205043.1 | NP_001191972.1 | 9 - 10 | 3133 | 94 | |
| NM_001205044.1 | NP_001191973.1 | 10 - 11 | 2806 | 94 | ||
| NM_021878.3 | NP_068678.1 | 10 - 11 | 2852 | 94 | ||
| GenBank mRNA | AK045214.1 | - | 9 - 10 | 2938 | 94 | |
| AK147226.1 | - | 9 - 10 | 3133 | 94 | ||
| AK164134.1 | - | 8 - 9 | 2457 | 94 | ||
| BC026056.1 | - | 9 - 10 | 3040 | 94 | ||
| BC050129.1 | - | 9 - 10 | 3054 | 94 | ||
| BC051010.1 | - | 9 - 10 | 3036 | 94 | ||
| BC052444.1 | - | 10 - 11 | 2723 | 6406875 | 94 | |
| BC060695.1 | - | 10 - 11 | 2723 | 6406875 | 94 | |
| BC115758.1 | - | 4 - 5 | 1737 | 94 | ||
| D31967.1 | - | 10 - 11 | 2742 | 94 | ||
More Information
Set Membership: |
|

Jarid2
Genomic Map