Assay Details
| Gene Symbol: Gabrb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038701.2 | NP_001033790.1 | 4 - 5 | 508 | 57 | |
| NM_008071.3 | NP_032097.1 | 4 - 5 | 560 | 57 | ||
| GenBank mRNA | AK039070.1 | - | 4 - 5 | 530 | 57 | |
| AK083051.1 | - | 4 - 5 | 572 | 57 | ||
| AK083107.1 | - | 4 - 5 | 509 | 57 | ||
| AK147247.1 | - | 4 - 5 | 514 | 57 | ||
| U14420.1 | - | 4 - 5 | 541 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gabrb3
Genomic Map