Assay Details
| Gene Symbol: Gabrb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038701.2 | NP_001033790.1 | 7 - 8 | 885 | 87 | |
| NM_008071.3 | NP_032097.1 | 7 - 8 | 937 | 87 | ||
| GenBank mRNA | AK039070.1 | - | 7 - 8 | 907 | 87 | |
| AK083051.1 | - | 7 - 8 | 949 | 87 | ||
| AK083107.1 | - | 7 - 8 | 886 | 87 | ||
| AK140718.1 | - | 3 - 4 | 479 | 87 | ||
| AK147247.1 | - | 7 - 8 | 891 | 87 | ||
More Information
Set Membership: |
|

Gabrb3
Genomic Map