Assay Details
| Gene Symbol: Siglech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178706.4 | NP_848821.2 | 3 - 4 | 868 | 72 | |
| GenBank mRNA | AK045719.1 | - | 4 - 5 | 498 | 72 | |
| AK080658.1 | - | 2 - 3 | 583 | 72 | ||
| AK139362.1 | - | 4 - 5 | 711 | 72 | ||
| BC098229.1 | - | 3 - 4 | 807 | 1430130 | 72 | |
| BC120501.1 | - | 3 - 4 | 810 | 40129424 | 72 | |
| BC144741.1 | - | 2 - 3 | 531 | 9053289 | 72 | |
| BC144742.1 | - | 3 - 4 | 810 | 9053290 | 72 | |
| BC144743.1 | - | 3 - 4 | 810 | 9053291 | 72 | |
| DQ157764.1 | - | 3 - 4 | 787 | 72 | ||
| DQ208935.1 | - | 3 - 4 | 813 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Siglech
Genomic Map