Assay Details
| Gene Symbol: St8sia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145838.1 | NP_665837.1 | 6 - 7 | 713 | 112 | |
| GenBank mRNA | AB059554.1 | - | 6 - 7 | 713 | 112 | |
| AK085105.1 | - | 7 - 8 | 850 | 112 | ||
| AK156557.1 | - | 6 - 7 | 1250 | 112 | ||
| AK172400.1 | - | 6 - 7 | 778 | 112 | ||
| BC115912.1 | - | 6 - 7 | 668 | 40104779 | 112 | |
| BC118063.1 | - | 6 - 7 | 668 | 40104780 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

St8sia6
Genomic Map