Assay Details
| Gene Symbol: Fam208b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134063.3 | NP_598824.3 | 9 - 10 | 1042 | 123 | |
| GenBank mRNA | AK138119.1 | - | 9 - 10 | 1043 | 123 | |
| AK220437.1 | - | 9 - 10 | 1030 | 123 | ||
| BC075685.1 | - | 7 - 8 | 518 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam208b
Genomic Map