Assay Details
| Gene Symbol: Pik3r5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177320.2 | NP_796294.2 | 5 - 6 | 563 | 118 | |
| GenBank mRNA | AK029201.1 | - | 3 - 4 | 231 | 118 | |
| AK089483.1 | - | 5 - 6 | 568 | 118 | ||
| AK149847.1 | - | 5 - 6 | 531 | 118 | ||
| AK154643.1 | - | 5 - 6 | 541 | 119 | ||
| AK171279.1 | - | 5 - 6 | 564 | 118 | ||
| AY156924.1 | - | 4 - 5 | 412 | 118 | ||
| BC128076.1 | - | 4 - 5 | 412 | 40060132 | 118 | |
| BC128077.1 | - | 4 - 5 | 412 | 40060133 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r5
Genomic Map