Assay Details
| Gene Symbol: Alox8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009661.4 | NP_033791.1 | 9 - 10 | 1330 | 85 | |
| GenBank mRNA | AK028724.1 | - | 9 - 10 | 1326 | 85 | |
| AK158553.1 | - | 7 - 8 | 1204 | 85 | ||
| BC015253.1 | - | 9 - 10 | 1330 | 4221815 | 85 | |
| U93277.1 | - | 9 - 10 | 1296 | 85 | ||
| Y14696.1 | - | 9 - 10 | 1309 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alox8
Genomic Map