Assay Details
| Gene Symbol: Arhgef15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177566.3 | NP_808234.2 | 3 - 4 | 1076 | 73 | |
| GenBank mRNA | BC089365.1 | - | 3 - 4 | 1076 | 6844583 | 73 |
| BC116343.1 | - | 3 - 4 | 1052 | 40098168 | 73 | |
| BC116344.1 | - | 3 - 4 | 1052 | 40098169 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef15
Genomic Map