Assay Details
| Gene Symbol: Ovol2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152947.2 | NP_694455.2 | 1 - 2 | 60 | 65 | |
| GenBank mRNA | AB101296.1 | - | 1 - 2 | 394 | 65 | |
| AY090538.1 | - | 1 - 2 | 60 | 65 | ||
| BY098454.1 | - | 1 - 2 | 60 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB101296.1, AY090538.1, NM_152947.2 transcripts.
Set Membership: |
|

Ovol2
Genomic Map