Assay Details
| Gene Symbol: Ovol2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026924.3 | NP_081200.2 | 2 - 3 | 536 | 94 | |
| NM_152947.2 | NP_694455.2 | 2 - 3 | 277 | 94 | ||
| GenBank mRNA | AB101294.1 | - | 2 - 3 | 439 | 94 | |
| AB101295.1 | - | 1 - 2 | 481 | 94 | ||
| AB101296.1 | - | 2 - 3 | 611 | 94 | ||
| AK007360.1 | - | 1 - 2 | 162 | 94 | ||
| AK167697.1 | - | 2 - 3 | 527 | 94 | ||
| AY090537.1 | - | 2 - 3 | 347 | 94 | ||
| AY090538.1 | - | 2 - 3 | 277 | 94 | ||
| BC031771.1 | - | 2 - 3 | 536 | 4219296 | 94 | |
| BC057210.1 | - | 2 - 3 | 557 | 5329625 | 94 | |
| BC094445.1 | - | 2 - 3 | 548 | 4981833 | 94 | |
| BY098454.1 | - | 2 - 3 | 277 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB101295.1, AK007360.1 transcripts.
Set Membership: |
|

Ovol2
Genomic Map