Assay Details
| Gene Symbol: Rin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161365.1 | NP_001154837.1 | 6 - 7 | 2119 | 85 | |
| NM_177620.4 | NP_808288.2 | 6 - 7 | 2230 | 85 | ||
| GenBank mRNA | AK032205.1 | - | 6 - 7 | 2231 | 85 | |
| AK154506.1 | - | 6 - 7 | 2255 | 85 | ||
| AK154522.1 | - | 6 - 7 | 2034 | 85 | ||
| AK154963.1 | - | 6 - 7 | 2120 | 85 | ||
| AK166196.1 | - | 6 - 7 | 2076 | 85 | ||
| AK171301.1 | - | 6 - 7 | 2044 | 85 | ||
| AK171311.1 | - | 6 - 7 | 2035 | 85 | ||
| AY265356.1 | - | 6 - 7 | 2216 | 85 | ||
| BC137907.1 | - | 6 - 7 | 2158 | 8860926 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rin3
Genomic Map