Assay Details
| Gene Symbol: Wwc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170779.1 | NP_740749.1 | 13 - 14 | 2003 | 86 | |
| GenBank mRNA | AB271907.1 | - | 13 - 14 | 1973 | 86 | |
| AK220259.1 | - | 2374 | 86 | |||
| BC006733.1 | - | 10 - 11 | 1592 | 86 | ||
| BC017638.1 | - | 6 - 7 | 1129 | 86 | ||
| BC037006.1 | - | 5 - 6 | 851 | 5116674 | 86 | |
| DQ256090.1 | - | 13 - 14 | 2003 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wwc1
Genomic Map