Assay Details
| Gene Symbol: Wwc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170779.1 | NP_740749.1 | 14 - 15 | 2094 | 69 | |
| GenBank mRNA | AB271907.1 | - | 14 - 15 | 2064 | 69 | |
| AK220259.1 | - | 2465 | 69 | |||
| BC006733.1 | - | 11 - 12 | 1683 | 69 | ||
| BC017638.1 | - | 7 - 8 | 1220 | 69 | ||
| BC037006.1 | - | 6 - 7 | 942 | 5116674 | 69 | |
| DQ256090.1 | - | 14 - 15 | 2094 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wwc1
Genomic Map