Assay Details
| Gene Symbol: Usp6nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080548.1 | NP_001074017.1 | 2 - 3 | 292 | 82 | |
| GenBank mRNA | AK084045.1 | - | 2 - 3 | 115 | 82 | |
| AK129037.1 | - | 2 - 3 | 144 | 82 | ||
| AK155073.1 | - | 2 - 3 | 293 | 82 | ||
| BE949333.1 | - | 2 - 3 | 381 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK155073.1, NM_001080548.1 transcripts.
Set Membership: |
|

Usp6nl
Genomic Map