Assay Details
| Gene Symbol: Usp6nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080548.1 | NP_001074017.1 | 8 - 9 | 779 | 84 | |
| NM_181399.3 | NP_852064.2 | 7 - 8 | 618 | 84 | ||
| GenBank mRNA | AK129037.1 | - | 8 - 9 | 631 | 84 | |
| AK155073.1 | - | 8 - 9 | 780 | 84 | ||
| BC051184.1 | - | 7 - 8 | 621 | 84 | ||
| BQ938063.1 | - | 7 - 8 | 618 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp6nl
Genomic Map