Assay Details
| Gene Symbol: Bend7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190400.1 | NP_001177329.1 | 4 - 5 | 972 | 96 | |
| NM_178663.4 | NP_848778.1 | 4 - 5 | 972 | 96 | ||
| GenBank mRNA | AK030428.1 | - | 4 - 5 | 659 | 96 | |
| AK042822.1 | - | 4 - 5 | 970 | 96 | ||
| AK053896.1 | - | 4 - 5 | 974 | 96 | ||
| BC096430.1 | - | 4 - 5 | 644 | 5041388 | 96 | |
| BC119370.1 | - | 4 - 5 | 611 | 8734119 | 96 | |
| BC144864.1 | - | 4 - 5 | 611 | 9053412 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bend7
Genomic Map