Assay Details
| Gene Symbol: Bend7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190400.1 | NP_001177329.1 | 6 - 7 | 1471 | 77 | |
| NM_178663.4 | NP_848778.1 | 6 - 7 | 1471 | 77 | ||
| GenBank mRNA | AK030428.1 | - | 6 - 7 | 1158 | 77 | |
| AK042822.1 | - | 6 - 7 | 1469 | 77 | ||
| AK053896.1 | - | 6 - 7 | 1473 | 77 | ||
| BC056476.1 | - | 3 - 4 | 515 | 77 | ||
| BC096430.1 | - | 6 - 7 | 1143 | 5041388 | 77 | |
| BC119370.1 | - | 6 - 7 | 1110 | 8734119 | 77 | |
| BC144864.1 | - | 6 - 7 | 1110 | 9053412 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bend7
Genomic Map