Assay Details
| Gene Symbol: Eml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043335.1 | NP_001036800.1 | 15 - 16 | 2127 | 65 | |
| NM_001043336.2 | NP_001036801.1 | 15 - 16 | 1744 | 65 | ||
| NM_001286346.1 | NP_001273275.1 | 15 - 16 | 1881 | 65 | ||
| NM_001286347.1 | NP_001273276.1 | 16 - 17 | 1795 | 65 | ||
| GenBank mRNA | AK044662.1 | - | 15 - 16 | 1866 | 65 | |
| BC038273.1 | - | 3 - 4 | 392 | 65 | ||
| BC053094.1 | - | 15 - 16 | 1961 | 6400458 | 65 | |
| BC059839.1 | - | 15 - 16 | 1722 | 6417403 | 65 | |
| BC079582.1 | - | 15 - 16 | 1780 | 5702743 | 65 | |
| BC098329.1 | - | 14 - 15 | 1524 | 65 | ||
| BC125289.1 | - | 16 - 17 | 1766 | 40129804 | 65 | |
| BC144757.1 | - | 16 - 17 | 1766 | 65 | ||
| BC150975.1 | - | 15 - 16 | 1881 | 9087888 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eml1
Genomic Map