Assay Details
| Gene Symbol: Eml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043335.1 | NP_001036800.1 | 19 - 20 | 2468 | 86 | |
| NM_001043336.2 | NP_001036801.1 | 19 - 20 | 2085 | 86 | ||
| NM_001286346.1 | NP_001273275.1 | 19 - 20 | 2222 | 86 | ||
| NM_001286347.1 | NP_001273276.1 | 20 - 21 | 2136 | 86 | ||
| GenBank mRNA | AK044662.1 | - | 19 - 20 | 2196 | 86 | |
| BC038273.1 | - | 7 - 8 | 733 | 86 | ||
| BC053094.1 | - | 19 - 20 | 2302 | 6400458 | 86 | |
| BC059839.1 | - | 19 - 20 | 2063 | 6417403 | 86 | |
| BC079582.1 | - | 19 - 20 | 2121 | 5702743 | 86 | |
| BC098329.1 | - | 18 - 19 | 1865 | 86 | ||
| BC125289.1 | - | 20 - 21 | 2107 | 40129804 | 86 | |
| BC144757.1 | - | 20 - 21 | 2107 | 86 | ||
| BC150975.1 | - | 19 - 20 | 2222 | 9087888 | 86 | |
| KJ734705.1 | - | 1 - 2 | 85 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eml1
Genomic Map