Assay Details
| Gene Symbol: Gm16532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134752.1 | NP_001128224.1 | 3 - 4 | 384 | 81 | |
| GenBank mRNA | AK046418.1 | - | 3 - 4 | 385 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P217360 | AK046418.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P217360 | NM_001134752.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P217360 | AK046418.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P217360 | NM_001134752.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6431 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650908 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574888 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149440 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 893120 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966149 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 751411 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240435 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562324 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 721625 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22307 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154322 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P217360 | AK046418.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P217360 | NM_001134752.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P217360 | AK046418.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P217360 | NM_001134752.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06062260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06237718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06307165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06351990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05990820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06043298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06587172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06229004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06307163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955388 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10549864 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549865 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459717_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549862 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700000886.mm.1 | TC0700000079.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000886.mm.1 | NM_001134752.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000886.mm.1 | NM_001134752.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000887.mm.1 | TC0700000079.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000887.mm.1 | NM_001134752.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000887.mm.1 | NM_001134752.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 408997 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 101500110 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245436 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101500110 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101500110 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245436 | - | - |

Gm16532
Genomic Map