Assay Details
| Gene Symbol: Ddx31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033294.3 | NP_001028466.2 | 2 - 3 | 710 | 80 | |
| GenBank mRNA | AK030888.1 | - | 2 - 3 | 657 | 80 | |
| AK144739.1 | - | 2 - 3 | 423 | 80 | ||
| AK166327.1 | - | 2 - 3 | 80 | |||
| BC066017.1 | - | 2 - 3 | 347 | 6401415 | 80 | |
| BC132351.1 | - | 2 - 3 | 400 | 40130628 | 80 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ddx31
Genomic Map