Assay Details
| Gene Symbol: Acsl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033600.1 | NP_001028772.1 | 11 - 12 | 1609 | 73 | |
| NM_019477.3 | NP_062350.3 | 11 - 12 | 1612 | 73 | ||
| NM_207625.2 | NP_997508.1 | 11 - 12 | 1902 | 73 | ||
| GenBank mRNA | AB033885.1 | - | 11 - 12 | 1421 | 73 | |
| AB033886.1 | - | 11 - 12 | 1711 | 73 | ||
| AB033887.1 | - | 11 - 12 | 1759 | 73 | ||
| AJ243502.1 | - | 11 - 12 | 1457 | 73 | ||
| AK054387.1 | - | 11 - 12 | 1582 | 73 | ||
| BC016416.1 | - | 11 - 12 | 1483 | 3157957 | 73 | |
| BC058663.1 | - | 11 - 12 | 1698 | 6409748 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acsl4
Genomic Map