Assay Details
| Gene Symbol: Acsl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033600.1 | NP_001028772.1 | 13 - 14 | 1869 | 118 | |
| NM_019477.3 | NP_062350.3 | 13 - 14 | 1872 | 118 | ||
| NM_207625.2 | NP_997508.1 | 13 - 14 | 2162 | 118 | ||
| GenBank mRNA | AJ243502.1 | - | 13 - 14 | 1717 | 118 | |
| AK054387.1 | - | 13 - 14 | 1842 | 118 | ||
| BC016416.1 | - | 13 - 14 | 1743 | 3157957 | 118 | |
| BC058663.1 | - | 13 - 14 | 1958 | 6409748 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acsl4
Genomic Map