Assay Details
| Gene Symbol: Acsl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033600.1 | NP_001028772.1 | 4 - 5 | 695 | 81 | |
| NM_019477.3 | NP_062350.3 | 4 - 5 | 698 | 81 | ||
| NM_207625.2 | NP_997508.1 | 4 - 5 | 988 | 81 | ||
| GenBank mRNA | AB033885.1 | - | 4 - 5 | 507 | 81 | |
| AB033886.1 | - | 4 - 5 | 797 | 81 | ||
| AB033887.1 | - | 4 - 5 | 845 | 81 | ||
| AJ243502.1 | - | 4 - 5 | 543 | 81 | ||
| AK054387.1 | - | 4 - 5 | 668 | 81 | ||
| BC016416.1 | - | 4 - 5 | 569 | 3157957 | 81 | |
| BC058663.1 | - | 4 - 5 | 784 | 6409748 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acsl4
Genomic Map