Assay Details
| Gene Symbol: Acsl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033600.1 | NP_001028772.1 | 6 - 7 | 947 | 117 | |
| NM_019477.3 | NP_062350.3 | 6 - 7 | 950 | 117 | ||
| NM_207625.2 | NP_997508.1 | 6 - 7 | 1240 | 117 | ||
| GenBank mRNA | AB033885.1 | - | 6 - 7 | 759 | 117 | |
| AB033886.1 | - | 6 - 7 | 1049 | 117 | ||
| AB033887.1 | - | 6 - 7 | 1097 | 117 | ||
| AJ243502.1 | - | 6 - 7 | 795 | 117 | ||
| AK054387.1 | - | 6 - 7 | 920 | 117 | ||
| BC016416.1 | - | 6 - 7 | 821 | 3157957 | 117 | |
| BC058663.1 | - | 6 - 7 | 1036 | 6409748 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acsl4
Genomic Map