Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200008449.mm.1 |
NM_001081457.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008449.mm.1 |
NM_001081458.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008449.mm.1 |
NM_001135001.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008449.mm.1 |
NM_001135001.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008449.mm.1 |
NM_012023.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008449.mm.1 |
TC1200001207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008455.mm.1 |
NM_012023.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008455.mm.1 |
TC1200001207.mm.1 |
MTA TC |