Assay Details
| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028261.1 | 13 - 14 | 2001 | 67 | ||
| GenBank mRNA | AF498294.1 | - | 3 - 4 | 778 | 67 | |
| AK017440.1 | - | 3 - 4 | 730 | 67 | ||
| EU434919.1 | - | 18 - 19 | 1640 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rian
Genomic Map