Assay Details
| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028261.1 | 16 - 17 | 2323 | 76 | ||
| GenBank mRNA | AF498294.1 | - | 6 - 7 | 1100 | 76 | |
| AF498295.1 | - | 1 - 2 | 113 | 76 | ||
| AK017440.1 | - | 6 - 7 | 1052 | 76 | ||
| EU434919.1 | - | 21 - 22 | 1962 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rian
Genomic Map