Assay Details
| Gene Symbol: Supt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009297.2 | NP_033323.2 | 10 - 11 | 1395 | 88 | |
| GenBank mRNA | AK170533.1 | - | 10 - 11 | 1394 | 88 | |
| BC072657.1 | - | 10 - 11 | 1315 | 30536338 | 88 | |
| U40375.1 | - | 10 - 11 | 1393 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt6
Genomic Map