Assay Details
| Gene Symbol: Sprr2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003548.1 | 1 - 2 | 41 | 71 | ||
| GenBank mRNA | AY158991.1 | - | 1 - 2 | 41 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small proline-rich protein 2F | Sprr2f | -7.5kb | Sense | Chr. 3 - 92365187 - 92366442 on NCBI Build GRCm38 | |
| Downstream | small proline-rich protein 2H | Sprr2h | AI661967 | 10.5kb | Sense | Chr. 3 - 92385685 - 92387324 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY158991.1 transcripts.
Set Membership: |
|

Sprr2g
Genomic Map