Assay Details
| Gene Symbol: Mpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164733.1 | NP_001158205.1 | 5 - 6 | 701 | 92 | |
| NM_001164734.1 | NP_001158206.1 | 5 - 6 | 711 | 92 | ||
| NM_019939.2 | NP_064323.2 | 5 - 6 | 711 | 92 | ||
| GenBank mRNA | AF161181.1 | - | 5 - 6 | 575 | 92 | |
| AF199009.1 | - | 5 - 6 | 707 | 92 | ||
| AF199010.1 | - | 5 - 6 | 707 | 92 | ||
| AK144434.1 | - | 5 - 6 | 712 | 92 | ||
| AK164549.1 | - | 5 - 6 | 702 | 92 | ||
| BC042868.2 | - | 5 - 6 | 658 | 92 | ||
| BC046421.1 | - | 5 - 6 | 658 | 92 | ||
| BC069908.1 | - | 5 - 6 | 700 | 92 | ||
| BC106140.1 | - | 5 - 6 | 658 | 92 | ||
| BC138666.1 | - | 5 - 6 | 569 | 8861687 | 92 | |
| BC145363.1 | - | 5 - 6 | 569 | 9053921 | 92 | |
| BC145364.1 | - | 5 - 6 | 559 | 9053922 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpp6
Genomic Map