Assay Details
| Gene Symbol: Arhgap21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081364.3 | NP_001074833.3 | 16 - 17 | 3924 | 113 | |
| NM_001128084.2 | NP_001121556.2 | 17 - 18 | 3954 | 113 | ||
| GenBank mRNA | AK122508.1 | - | 1565 | 113 | ||
| BC076629.1 | - | 16 - 17 | 3924 | 30639338 | 113 | |
| BC150741.1 | - | 17 - 18 | 3883 | 9087652 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap21
Genomic Map