Assay Details
| Gene Symbol: Arhgap21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081364.3 | NP_001074833.3 | 19 - 20 | 4170 | 101 | |
| NM_001128084.2 | NP_001121556.2 | 20 - 21 | 4200 | 101 | ||
| GenBank mRNA | AK122508.1 | - | 1811 | 101 | ||
| BC076629.1 | - | 19 - 20 | 4170 | 30639338 | 101 | |
| BC150741.1 | - | 20 - 21 | 4129 | 9087652 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap21
Genomic Map