Assay Details
| Gene Symbol: Arhgap21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081364.3 | NP_001074833.3 | 21 - 22 | 4336 | 100 | |
| NM_001128084.2 | NP_001121556.2 | 22 - 23 | 4366 | 100 | ||
| GenBank mRNA | AK122508.1 | - | 1977 | 100 | ||
| BC076629.1 | - | 21 - 22 | 4336 | 30639338 | 100 | |
| BC150741.1 | - | 22 - 23 | 4295 | 9087652 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap21
Genomic Map