Assay Details
| Gene Symbol: Tax1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025816.3 | NP_080092.2 | 8 - 9 | 1166 | 78 | |
| GenBank mRNA | AK004574.1 | - | 8 - 9 | 1148 | 78 | |
| AK146076.1 | - | 8 - 9 | 1146 | 78 | ||
| BC014798.1 | - | 8 - 9 | 1087 | 3962810 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tax1bp1
Genomic Map