Assay Details
| Gene Symbol: Tax1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025816.3 | NP_080092.2 | 9 - 10 | 1396 | 107 | |
| GenBank mRNA | AK004574.1 | - | 9 - 10 | 1378 | 107 | |
| AK146076.1 | - | 9 - 10 | 1376 | 107 | ||
| BC014798.1 | - | 9 - 10 | 1317 | 3962810 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tax1bp1
Genomic Map