Assay Details
| Gene Symbol: Skap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018773.2 | NP_061243.1 | 7 - 8 | 748 | 64 | |
| GenBank mRNA | AB014485.1 | - | 7 - 8 | 665 | 64 | |
| AF051324.1 | - | 7 - 8 | 663 | 64 | ||
| AK076000.1 | - | 7 - 8 | 693 | 64 | ||
| AK144289.1 | - | 7 - 8 | 685 | 64 | ||
| BC003711.1 | - | 7 - 8 | 748 | 3599914 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Skap2
Genomic Map