Assay Details
| Gene Symbol: Crhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009953.4 | NP_034083.2 | 4 - 5 | 547 | 63 | |
| GenBank mRNA | AK085854.1 | - | 4 - 5 | 548 | 63 | |
| AK142261.1 | - | 4 - 5 | 497 | 63 | ||
| U19939.1 | - | 4 - 5 | 503 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crhr2
Genomic Map