Assay Details
| Gene Symbol: Crhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288618.1 | NP_001275547.1 | 5 - 6 | 661 | 67 | |
| NM_001288619.1 | NP_001275548.1 | 4 - 5 | 568 | 67 | ||
| NM_009953.4 | NP_034083.2 | 5 - 6 | 658 | 67 | ||
| GenBank mRNA | AK085854.1 | - | 5 - 6 | 659 | 67 | |
| AK142261.1 | - | 5 - 6 | 608 | 67 | ||
| AY445512.1 | - | 4 - 5 | 568 | 67 | ||
| BC137592.1 | - | 5 - 6 | 544 | 8860611 | 67 | |
| CJ065172.1 | - | 5 - 6 | 611 | 67 | ||
| FJ548565.1 | - | 5 - 6 | 486 | 67 | ||
| U17858.1 | - | 5 - 6 | 486 | 67 | ||
| U19939.1 | - | 5 - 6 | 614 | 67 | ||
| U21729.1 | - | 5 - 6 | 612 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crhr2
Genomic Map