Assay Details
| Gene Symbol: Inmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009349.3 | NP_033375.1 | 1 - 2 | 156 | 75 | |
| GenBank mRNA | AK002281.1 | - | 1 - 2 | 204 | 75 | |
| AK013010.1 | - | 1 - 2 | 193 | 75 | ||
| BC013518.1 | - | 1 - 2 | 182 | 4236077 | 75 | |
| M88694.1 | - | 1 - 2 | 157 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Inmt
Genomic Map