Assay Details
| Gene Symbol: Ghrhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003685.3 | NP_001003685.2 | 5 - 6 | 555 | 73 | |
| GenBank mRNA | AK030504.1 | - | 5 - 6 | 509 | 73 | |
| BC120748.1 | - | 5 - 6 | 485 | 40129671 | 73 | |
| BC120774.1 | - | 5 - 6 | 485 | 40129697 | 73 | |
| BC144892.1 | - | 5 - 6 | 485 | 73 | ||
| L07379.1 | - | 5 - 6 | 488 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ghrhr
Genomic Map