Assay Details
| Gene Symbol: Ghrhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003685.3 | NP_001003685.2 | 8 - 9 | 904 | 102 | |
| GenBank mRNA | AK030504.1 | - | 8 - 9 | 858 | 102 | |
| BC120748.1 | - | 8 - 9 | 834 | 40129671 | 102 | |
| BC120774.1 | - | 8 - 9 | 834 | 40129697 | 102 | |
| BC144892.1 | - | 8 - 9 | 834 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ghrhr
Genomic Map