Assay Details
| Gene Symbol: Gars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180678.3 | NP_851009.2 | 6 - 7 | 745 | 78 | |
| GenBank mRNA | AK012758.2 | - | 6 - 7 | 740 | 78 | |
| AK146238.1 | - | 6 - 7 | 722 | 78 | ||
| AK165857.1 | - | 6 - 7 | 693 | 78 | ||
| AK211890.1 | - | 2 - 3 | 120 | 78 | ||
| BC021747.1 | - | 6 - 7 | 726 | 4192628 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gars
Genomic Map