Assay Details
| Gene Symbol: Plekha8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001335.2 | NP_001001335.1 | 2 - 3 | 390 | 67 | |
| NM_001164361.1 | NP_001157833.1 | 2 - 3 | 485 | 67 | ||
| GenBank mRNA | AK078040.1 | - | 2 - 3 | 485 | 67 | |
| AK170592.1 | - | 2 - 3 | 252 | 67 | ||
| BC052360.1 | - | 2 - 3 | 390 | 6704789 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC052360.1, NM_001001335.2 transcripts.
Set Membership: |
|

Plekha8
Genomic Map