Assay Details
| Gene Symbol: Surf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271724.1 | NP_001258653.2 | 8 - 9 | 1015 | 94 | |
| NM_013677.2 | NP_038705.2 | 8 - 9 | 1051 | 94 | ||
| NR_073420.1 | 8 - 9 | 1091 | 94 | |||
| GenBank mRNA | AK002469.1 | - | 8 - 9 | 865 | 94 | |
| AK154726.1 | - | 8 - 9 | 871 | 95 | ||
| BC004755.1 | - | 8 - 9 | 1051 | 3584373 | 94 | |
| BC052500.1 | - | 8 - 9 | 1038 | 5255619 | 94 | |
| CB524655.1 | - | 3 - 4 | 321 | 94 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081132 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081131 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | AK208539.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | BC004755.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | BC052500.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | CB524655.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | AK208539.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | BC004755.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | BC052500.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | CB524655.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284271 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 145591 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178683 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | AK208539.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | BC004755.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | BC052500.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | CB524655.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | AK208539.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | BC004755.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | BC052500.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | CB524655.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | AK208539.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | BC004755.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | BC052500.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | CB524655.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | AK208539.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | BC004755.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | BC052500.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | CB524655.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081119 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178683 | - | - |
| Whole Mouse Genome G4122A | A_51_P332602 | AK208539.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332602 | BC004755.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332602 | BC052500.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332602 | CB524655.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332602 | AK208539.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332602 | BC004755.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332602 | BC052500.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332602 | CB524655.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332602 | NM_013677.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116405 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178683 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01852378 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01852385 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01852382 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01852373 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081132 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01582440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01582397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01582441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21081145 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10481145 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470323 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481146 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481136 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481137 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481141 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481139 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481142 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481143 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481144 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481140 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10481138 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450561_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10481135 | - | - |
| Mouse Genome 430 2.0 Array | 1450561_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450561_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20930_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20930_PM_at | - | - |
| Mouse Expression Set 430 | 1450561_a_at | - | - |
| Mouse Expression Set 430 | 1450561_PM_a_at | - | - |
| Murine Genome U74v2 Set | 101031_PM_at | - | - |
| Murine Genome U74v2 Set | 101031_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450561_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450561_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450561_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450561_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5130048 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5130048 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005130048 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2657141 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2657141 | - | - |
| mouse-6 v1.1 (Illumina) | scl19516.4.19_7-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5130048 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl19516.4.19_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5130048 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2657141 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2883561 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2883561 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2657141 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl19516.4.19_7-S | - | - |

Surf1
Genomic Map