Assay Details
| Gene Symbol: Ddx46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282055.1 | NP_001268984.1 | 12 - 13 | 1723 | 127 | |
| GenBank mRNA | BC026492.1 | - | 5 - 6 | 668 | 4505095 | 127 |
| BC092240.1 | - | 11 - 12 | 1634 | 30662573 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx46
Genomic Map